3-(4-Methoxyphenyl)isoxazole 95%

Supplier: Apollo Scientific

OR932210-1G OR932210-250MG
APOSOR932210-1GEA 647 EUR
APOSOR932210-1G APOSOR932210-250MG
3-(4-Methoxyphenyl)isoxazole 95%
3-(4-Methoxyphenyl)isoxazole
Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08446386
CAS Number: 61428-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR