(2-Chloro-thiazol-5-ylmethyl)-ethyl-amine 95%

Supplier: Apollo Scientific

Synonyms: N-((2-Chlorothiazol-5-yl)methyl)ethanamine

OR932883-1G OR932883-500MG
APOSOR932883-1GEA 829 EUR
APOSOR932883-1G APOSOR932883-500MG
(2-Chloro-thiazol-5-ylmethyl)-ethyl-amine 95%
(2-Chloro-thiazol-5-ylmethyl)-ethyl-amine
Formula: C₆H₉ClN₂S
MW: 176,667 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18837199
CAS Number: 120740-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR