2-P-Tolyloxymethyl-oxirane 95%

Supplier: Apollo Scientific

Synonyms: 2-((p-Tolyloxy)methyl)oxirane

OR932992-1G OR932992-250MG
APOSOR932992-1GEA 437 EUR
APOSOR932992-1G APOSOR932992-250MG
2-P-Tolyloxymethyl-oxirane 95%
2-P-Tolyloxymethyl-oxirane
Formula: C₁₀H₁₂O₂
MW: 164,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046991
CAS Number: 2186-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR