2-Chloro-3-methylphenol 96%

Supplier: Apollo Scientific

OR933107-1G OR933107-5G
APOSOR933107-1GEA 132 EUR
APOSOR933107-1G APOSOR933107-5G
2-Chloro-3-methylphenol 96%
2-Chloro-3-methylphenol
Formula: C₇H₇ClO
MW: 142,583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036178
CAS Number: 608-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR