2-Chloro-5-(methoxymethyl)pyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR933592-250MG
OR933592-100MG
APOSOR933592-250MGEA
190
EUR
APOSOR933592-250MG
APOSOR933592-100MG
2-Chloro-5-(methoxymethyl)pyrimidine
2-Chloro-5-(methoxymethyl)pyrimidine
Formula:
C₆H₇ClN₂O MW: 158,586 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28385449 CAS Number: 1416366-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...