3-Aminocyclobutane-1-carbonitrile hydrochloride

Supplier: Apollo Scientific

OR934826-1G OR934826-500MG
APOSOR934826-1GEA 1270 EUR
APOSOR934826-1G APOSOR934826-500MG
3-Aminocyclobutane-1-carbonitrile hydrochloride
3-Aminocyclobutane-1-carbonitrile hydrochloride
Formula: C₅H₈N₂·ClH
MW: 132,591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26959098
CAS Number: 1393180-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR