2-Cyanobenzyl acetate 95%
Supplier: Apollo Scientific
OR937471-10G
OR937471-1G
OR937471-5G
APOSOR937471-10GEA
109
EUR
APOSOR937471-10G
APOSOR937471-1G
APOSOR937471-5G
2-Cyanobenzyl acetate 95%
2-Cyanobenzyl acetate
Formula:
C₁₀H₉NO₂ MW: 175,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20265196 CAS Number: 38866-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...