5-Cyano-2-cyclopentyloxypyridine 97%

Supplier: Apollo Scientific

OR937608-1G OR937608-250MG OR937608-5G
APOSOR937608-1GEA 226 EUR
APOSOR937608-1G APOSOR937608-250MG APOSOR937608-5G
5-Cyano-2-cyclopentyloxypyridine 97%
5-Cyano-2-cyclopentyloxypyridine
Formula: C₁₁H₁₂N₂O
MW: 188,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09940721
CAS Number: 942938-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR