Cycloheptanecarbaldehyde 95%

Supplier: Apollo Scientific

OR938281-100MG OR938281-250MG OR938281-500MG
APOSOR938281-100MGEA 203 EUR
APOSOR938281-100MG APOSOR938281-250MG APOSOR938281-500MG
Cycloheptanecarbaldehyde 95%
Cycloheptanecarbaldehyde
Formula: C₈H₁₄O
MW: 126.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412495
CAS Number: 4277-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR