3-(Cyclopropyl)benzoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Cyclopropylbenzoic acid
OR938340-1G
OR938340-250MG
OR938340-5G
APOSOR938340-1GEA
111
EUR
APOSOR938340-1G
APOSOR938340-250MG
APOSOR938340-5G
3-(Cyclopropyl)benzoic acid
3-(Cyclopropyl)benzoic acid
Formula:
C₁₀H₁₀O₂ MW: 162.185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00626842 CAS Number: 1129-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...