[(3S)-1,4-Dioxaspiro[4.5]decan-3-yl]methanol 95%

Supplier: Apollo Scientific

OR938463-1G OR938463-5G
APOSOR938463-1GEA 161 EUR
APOSOR938463-1G APOSOR938463-5G
[(3S)-1,4-Dioxaspiro[4.5]decan-3-yl]methanol 95%
[(3S)-1,4-Dioxaspiro[4.5]decan-3-yl]methanol
Formula: C₉H₁₆O₃
MW: 172,222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276891
CAS Number: 95335-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR