trans-3-(Trimethylsilyl)allyl alcohol 95%

Supplier: Apollo Scientific

OR938640-1G OR938640-5G
APOSOR938640-1GEA 306 EUR
APOSOR938640-1G APOSOR938640-5G
trans-3-(Trimethylsilyl)allyl alcohol 95%
trans-3-(Trimethylsilyl)allyl alcohol
Formula: C₆H₁₄OSi
MW: 130,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077889
CAS Number: 18146-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR