1-(tert-Butyldimethylsilyloxy)-2-propanone 97%

Supplier: Apollo Scientific

Synonyms: 1-((tert-Butyldimethylsilyl)oxy)propan-2-one

OR938890-25G
APOSOR938890-25GEA 139 EUR
APOSOR938890-25G
1-(tert-Butyldimethylsilyloxy)-2-propanone 97%
1-(tert-Butyldimethylsilyloxy)-2-propanone
Formula: C₉H₂₀O₂Si
MW: 188,339 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321330
CAS Number: 74685-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR