N-(2-Oxo-2-phenylethyl)acetamide 95%

Supplier: Apollo Scientific

OR938951-1G OR938951-5G
APOSOR938951-1GEA 328 EUR
APOSOR938951-1G APOSOR938951-5G
N-(2-Oxo-2-phenylethyl)acetamide 95%
N-(2-Oxo-2-phenylethyl)acetamide
Formula: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19313693
CAS Number: 1846-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR