2-Ethynylphenol 95%
Supplier: Apollo Scientific
OR939099-1G
OR939099-250MG
APOSOR939099-1GEA
322
EUR
APOSOR939099-1G
APOSOR939099-250MG
2-Ethynylphenol 95%
2-Ethynylphenol
Formula:
C₈H₆O MW: 118.133 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00078394 CAS Number: 5101-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...