5,6,7,8-Tetrahydronaphthalen-2-amine 95%

Supplier: Apollo Scientific

Synonyms: 5,6,7,8-Tetrahydro-2-naphthylamine

OR939187-1G OR939187-5G
APOSOR939187-1GEA 132 EUR
APOSOR939187-1G APOSOR939187-5G
5,6,7,8-Tetrahydronaphthalen-2-amine 95%
5,6,7,8-Tetrahydronaphthalen-2-amine
Formula: C₁₀H₁₃N
MW: 147,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00130046
CAS Number: 2217-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR