6-Chlorobenzo[d]thiazol-2(3H)-one 95%

Supplier: Apollo Scientific

OR939213-1G OR939213-250MG
APOSOR939213-1GEA 82.9 EUR
APOSOR939213-1G APOSOR939213-250MG
6-Chlorobenzo[d]thiazol-2(3H)-one 95%
6-Chlorobenzo[d]thiazol-2(3H)-one
Formula: C₇H₄ClNOS
MW: 185,631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783782
CAS Number: 62266-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR