6-Methyl-2-(methylthio)-1H-pyrimidin-4-one 95%
Supplier: Apollo Scientific
OR939412-10G
OR939412-25G
APOSOR939412-10GEA
105
EUR
APOSOR939412-10G
APOSOR939412-25G
6-Methyl-2-(methylthio)-1H-pyrimidin-4-one 95%
6-Methyl-2-(methylthio)-1H-pyrimidin-4-one
Formula:
C₆H₈N₂OS MW: 156,206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01022058 CAS Number: 6328-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...