Methyl 3-oxocyclohexanecarboxylate 95%

Supplier: Apollo Scientific

OR939437-1G OR939437-250MG
APOSOR939437-1GEA 109 EUR
APOSOR939437-1G APOSOR939437-250MG
Methyl 3-oxocyclohexanecarboxylate 95%
Methyl 3-oxocyclohexanecarboxylate
Formula: C₈H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09031996
CAS Number: 13148-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR