(+)-Methyl (s)-3-hydroxyvalerate 95%

Supplier: Apollo Scientific

OR939606-1G OR939606-250MG
APOSOR939606-1GEA 180 EUR
APOSOR939606-1G APOSOR939606-250MG
(+)-Methyl (s)-3-hydroxyvalerate 95%
(+)-Methyl (s)-3-hydroxyvalerate
Formula: C₆H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00216722
CAS Number: 42558-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR