2-Chloro-3-hydroxybenzonitrile 95%

Supplier: Apollo Scientific

OR939696-1G OR939696-250MG
APOSOR939696-1GEA 198 EUR
APOSOR939696-1G APOSOR939696-250MG
2-Chloro-3-hydroxybenzonitrile 95%
2-Chloro-3-hydroxybenzonitrile
Formula: C₇H₄ClNO
MW: 153,566 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848763
CAS Number: 51786-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR