2-(3-Chlorophenyl)propanoic acid 95%

Supplier: Apollo Scientific

OR939790-10G OR939790-25G OR939790-5G
APOSOR939790-10GEA 328 EUR
APOSOR939790-10G APOSOR939790-25G APOSOR939790-5G
2-(3-Chlorophenyl)propanoic acid 95%
2-(3-Chlorophenyl)propanoic acid
Formula: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412454
CAS Number: 14161-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR