2-Isopropylbenzaldehyde 95%
Supplier: Apollo Scientific
OR939813-1G
OR939813-250MG
OR939813-5G
APOSOR939813-1GEA
256
EUR
APOSOR939813-1G
APOSOR939813-250MG
APOSOR939813-5G
2-Isopropylbenzaldehyde 95%
2-Isopropylbenzaldehyde
Formula:
C₁₀H₁₂O MW: 148,202 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06201222 CAS Number: 6502-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...