Ethyl 2,2-dichloro-3-oxobutanoate 95%
Supplier: Apollo Scientific
|
Warning
|
OR939947-250MG
OR939947-5G
APOSOR939947-250MGEA
364
EUR
APOSOR939947-250MG
APOSOR939947-5G
Ethyl 2,2-dichloro-3-oxobutanoate 95%
Ethyl 2,2-dichloro-3-oxobutanoate
Formula:
C₆H₈Cl₂O₃ MW: 199,032 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00129136 CAS Number: 6134-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...