3-(Diethylamino)-2,2-dimethylpropan-1-ol 95%
Supplier: Apollo Scientific
OR940572-1G
OR940572-5G
APOSOR940572-1GEA
0
EUR
APOSOR940572-1G
APOSOR940572-5G
3-(Diethylamino)-2,2-dimethylpropan-1-ol 95%
3-(Diethylamino)-2,2-dimethylpropan-1-ol
Formula:
C₉H₂₁NO MW: 159,269 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085710 CAS Number: 39067-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...