(S)-Piperidin-3-ylmethanol hydrochloride 95%
Supplier: Apollo Scientific
OR940701-1G
OR940701-250MG
APOSOR940701-1GEA
80
EUR
APOSOR940701-1G
APOSOR940701-250MG
(S)-Piperidin-3-ylmethanol hydrochloride 95%
(S)-Piperidin-3-ylmethanol hydrochloride
Formula:
C₆H₁₃NO·ClH MW: 151,634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14155681 CAS Number: 1125551-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...