Ethyl 1-aminocyclopentanecarboxylate 95%

Supplier: Apollo Scientific

OR940716-1G OR940716-250MG
APOSOR940716-1GEA 844 EUR
APOSOR940716-1G APOSOR940716-250MG
Ethyl 1-aminocyclopentanecarboxylate 95%
Ethyl 1-aminocyclopentanecarboxylate
Formula: C₈H₁₅NO₂
MW: 157,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663001
CAS Number: 1664-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR