3-Nitro-1H-pyrrolo[3,2-b]pyridine 95%
Supplier: Apollo Scientific
OR941059-1G
OR941059-250MG
APOSOR941059-1GEA
807
EUR
APOSOR941059-1G
APOSOR941059-250MG
3-Nitro-1H-pyrrolo[3,2-b]pyridine 95%
3-Nitro-1H-pyrrolo[3,2-b]pyridine
Formula:
C₇H₅N₃O₂ MW: 163,133 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846273 CAS Number: 23612-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...