Ethyl 5-hydroxypicolinate 95%
Supplier: Apollo Scientific
OR941184-1G
OR941184-250MG
APOSOR941184-1GEA
735
EUR
APOSOR941184-1G
APOSOR941184-250MG
Ethyl 5-hydroxypicolinate 95%
Ethyl 5-hydroxypicolinate
Formula:
C₈H₉NO₃ MW: 167,162 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11656385 CAS Number: 65275-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...