3-Acetamido-4-picoline 98%
Supplier: Apollo Scientific
Synonyms:
N-(4-Methylpyridin-3-yl)acetamide
OR941452-1G
APOSOR941452-1GEA
53.8
EUR
APOSOR941452-1G
3-Acetamido-4-picoline 98%
3-Acetamido-4-picoline
Formula:
C₈H₁₀N₂O MW: 150,178 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07776942 CAS Number: 52090-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...