Dimethyl-(5-methyl-pyridin-2-yl)-amine 95%
Supplier: Apollo Scientific
OR941719-1G
OR941719-250MG
APOSOR941719-1GEA
306
EUR
APOSOR941719-1G
APOSOR941719-250MG
Dimethyl-(5-methyl-pyridin-2-yl)-amine 95%
Dimethyl-(5-methyl-pyridin-2-yl)-amine
Formula:
C₈H₁₂N₂ MW: 136,194 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06411127 CAS Number: 147151-28-8 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...