5-Benzyl-1,3,4-oxadiazol-2-ylamine 95%

Supplier: Apollo Scientific

OR941940-1G OR941940-5G
APOSOR941940-1GEA 174 EUR
APOSOR941940-1G APOSOR941940-5G
5-Benzyl-1,3,4-oxadiazol-2-ylamine 95%
5-Benzyl-1,3,4-oxadiazol-2-ylamine
Formula: C₉H₉N₃O
MW: 175,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159750
CAS Number: 31803-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR