N-Ethylpyridin-3-amine 95%

Supplier: Apollo Scientific

OR942007-1G OR942007-250MG
APOSOR942007-1GEA 488 EUR
APOSOR942007-1G APOSOR942007-250MG
N-Ethylpyridin-3-amine 95%
N-Ethylpyridin-3-amine
Formula: C₇H₁₀N₂
MW: 122,168 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11140216
CAS Number: 32405-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR