(2S,6S)-2,6-Dimethylmorpholine

Supplier: Apollo Scientific
Warning

OR942218-1G OR942218-250MG
APOSOR942218-1GEA 252 EUR
APOSOR942218-1G APOSOR942218-250MG
(2S,6S)-2,6-Dimethylmorpholine
(2S,6S)-2,6-Dimethylmorpholine
Formula: C₆H₁₃NO
MW: 115.174 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18250271
CAS Number: 276252-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR