4-(Benzyloxy)pyridine 95%

Supplier: Apollo Scientific

OR942402-1G
APOSOR942402-1GEA 74.1 EUR
APOSOR942402-1G
4-(Benzyloxy)pyridine 95%
4-(Benzyloxy)pyridine
Formula: C₁₂H₁₁NO
MW: 185,222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08235112
CAS Number: 49826-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR