4-Methoxy-N,N-dimethylaniline 95%
Supplier: Apollo Scientific
OR942467-1G
OR942467-5G
APOSOR942467-1GEA
64
EUR
APOSOR942467-1G
APOSOR942467-5G
4-Methoxy-N,N-dimethylaniline 95%
4-Methoxy-N,N-dimethylaniline
Formula:
C₉H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD00155300 CAS Number: 701-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...