8-Chloroimidazo[1,2-a]pyridine 95%

Supplier: Apollo Scientific

OR942577-1G OR942577-250MG
APOSOR942577-1GEA 68.4 EUR
APOSOR942577-1G APOSOR942577-250MG
8-Chloroimidazo[1,2-a]pyridine 95%
8-Chloroimidazo[1,2-a]pyridine
Formula: C₇H₅ClN₂
MW: 152,581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658689
CAS Number: 1195251-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR