6-Bromo-3-pyridazinol 98%
Supplier: Apollo Scientific
Synonyms:
6-Bromopyridazin-3(2H)-one
OR943012-10G
OR943012-1G
OR943012-5G
APOSOR943012-10GEA
342
EUR
APOSOR943012-10G
APOSOR943012-1G
APOSOR943012-5G
6-Bromo-3-pyridazinol 98%
6-Bromo-3-pyridazinol
Formula:
C₄H₃BrN₂O MW: 174.983 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11223475 CAS Number: 51355-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...