2-(Azetidin-1-yl)ethanol 95%

Supplier: Apollo Scientific
Warning

OR943468-1G OR943468-250MG
APOSOR943468-1GEA 614 EUR
APOSOR943468-1G APOSOR943468-250MG
2-(Azetidin-1-yl)ethanol 95%
2-(Azetidin-1-yl)ethanol
Formula: C₅H₁₁NO
MW: 101,147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09743797
CAS Number: 67896-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR