2-Chloro-1-(2,3-dihydro-indol-1-yl)-ethanone 95%

Supplier: Apollo Scientific

Synonyms: 2-Chloro-1-(indolin-1-yl)ethanone

OR943541-1G OR943541-250MG
APOSOR943541-1GEA 219 EUR
APOSOR943541-1G APOSOR943541-250MG
2-Chloro-1-(2,3-dihydro-indol-1-yl)-ethanone 95%
2-Chloro-1-(2,3-dihydro-indol-1-yl)-ethanone
Formula: C₁₀H₁₀ClNO
MW: 195,646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00099469
CAS Number: 17133-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR