N-Methoxy-N,3-dimethylbutanamide 98%
Supplier: Apollo Scientific
OR943818-1G
OR943818-5G
APOSOR943818-1GEA
190
EUR
APOSOR943818-1G
APOSOR943818-5G
N-Methoxy-N,3-dimethylbutanamide 98%
N-Methoxy-N,3-dimethylbutanamide
Formula:
C₇H₁₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09859663 CAS Number: 147356-77-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...