8-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR943847-1G
OR943847-250MG
APOSOR943847-1GEA
807
EUR
APOSOR943847-1G
APOSOR943847-250MG
8-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 95%
8-Methylimidazo[1,2-a]pyridine-3-carbaldehyde
Formula:
C₉H₈N₂O MW: 160,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09994355 CAS Number: 175878-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...