2-Ethynyl-6-methoxypyridine 98%
Supplier: Apollo Scientific
OR943903-1G
OR943903-250MG
APOSOR943903-1GEA
806
EUR
APOSOR943903-1G
APOSOR943903-250MG
2-Ethynyl-6-methoxypyridine 98%
2-Ethynyl-6-methoxypyridine
Formula:
C₈H₇NO Storage Temperature: Ambient |
MDL Number:
MFCD11109683 CAS Number: 512197-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...