5-Chloro-1,3,4-thiadiazole-2-carboxamide
Supplier: Apollo Scientific
|
Warning
|
OR944093-1G
OR944093-250MG
APOSOR944093-1GEA
290
EUR
APOSOR944093-1G
APOSOR944093-250MG
5-Chloro-1,3,4-thiadiazole-2-carboxamide
5-Chloro-1,3,4-thiadiazole-2-carboxamide
Formula:
C₃H₂ClN₃OS MW: 163,585 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16659015 CAS Number: 64837-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...