Ethyl 3-butenoate 90%

Supplier: Apollo Scientific

Synonyms: Ethyl but-3-enoate

OR944678-1G OR944678-250MG
APOSOR944678-1GEA 698 EUR
APOSOR944678-1G APOSOR944678-250MG
Ethyl 3-butenoate 90%
Ethyl 3-butenoate
Formula: C₆H₁₀O₂
MW: 114,142 g/mol
Boiling Pt: 117 °C
Density: 0,904 g/cm³ (25 °C)
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD07779251
CAS Number: 1617-18-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR