6-Chloro-2,2-dimethyl-3,4-dihydro-1,4-benzoxazine 95%
Supplier: Apollo Scientific
Synonyms:
6-Chloro-2,2-dimethyl-3,4-dihydro-2H-benzo[b][1,4]oxazine
OR945251-1G
OR945251-250MG
APOSOR945251-1GEA
80
EUR
APOSOR945251-1G
APOSOR945251-250MG
6-Chloro-2,2-dimethyl-3,4-dihydro-1,4-benzoxazine 95%
6-Chloro-2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
Formula:
C₁₀H₁₂ClNO MW: 197,661 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14539604 CAS Number: 1216138-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...