1-Methyl-1H-2,1-benzothiazin-4(3H)-one 2,2-dioxide

Supplier: Apollo Scientific
Warning

OR9458-1G OR9458-250MG OR9458-5G
APOSOR9458-1GEA 68.3 EUR
APOSOR9458-1G APOSOR9458-250MG APOSOR9458-5G
1-Methyl-1H-2,1-benzothiazin-4(3H)-one 2,2-dioxide
1-Methyl-1H-2,1-benzothiazin-4(3H)-one 2,2-dioxide
Formula: C₉H₉NO₃S
Melting Pt: 117…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00067994
CAS Number: 7117-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR