3-Cyano-N-cyclopropylbenzamide 98%

Supplier: Apollo Scientific

OR946461-1G OR946461-5G
APOSOR946461-1GEA 219 EUR
APOSOR946461-1G APOSOR946461-5G
3-Cyano-N-cyclopropylbenzamide 98%
3-Cyano-N-cyclopropylbenzamide
Formula: C₁₁H₁₀N₂O
MW: 186,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09946575
CAS Number: 1016788-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR