1-(1-Isobutylpiperidin-4-yl)methanamine 95%

Supplier: Apollo Scientific

OR946554-1G OR946554-250MG
APOSOR946554-1GEA 647 EUR
APOSOR946554-1G APOSOR946554-250MG
1-(1-Isobutylpiperidin-4-yl)methanamine 95%
1-(1-Isobutylpiperidin-4-yl)methanamine
Formula: C₁₀H₂₂N₂
MW: 170,295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07365294
CAS Number: 258345-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR