3-(Isobutanoylamino)benzeneboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Isobutyramidobenzeneboronic acid
OR9466-1G
OR9466-5G
APOSOR9466-1GEA
183
EUR
APOSOR9466-1G
APOSOR9466-5G
3-(Isobutanoylamino)benzeneboronic acid 98%
3-(Isobutanoylamino)benzeneboronic acid
Formula:
C₁₀H₁₄BNO₃ MW: 207,034 g/mol Melting Pt: 275…277 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08235080 CAS Number: 874459-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...